ENA413171885 Enamine

4-chloro-2-(3-methyl-1,2,4-oxadiazol-5-yl)phenol

  • MDL number MFCD18898356
  • Cas Number : 1343260-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156447
Availability details for
Quantity
  • Availability
  • Add to Cart