ENA413171891 Enamine

6-amino-1-(2,2-difluoroethyl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12807524
  • Cas Number : 1179946-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156457
Availability details for
Quantity
  • Availability
  • Add to Cart