ENA413171898 Enamine

4-(methoxymethyl)-1H-imidazole

  • MDL number MFCD19218079
  • Cas Number : 875155-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156497
Availability details for
Quantity
  • Availability
  • Add to Cart