ENA413171906 Enamine

phenyl N-(cyclopropylmethyl)carbamate

  • MDL number MFCD11100385
  • Cas Number : 66849-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156522

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156522
Availability details for
Quantity
  • Availability
  • Add to Cart