ENA413171907 Enamine

2',6'-dioxospiro[adamantane-2,4'-piperidine]-3',5'-dicarbonitrile

  • Cas Number : 405889-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156523

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156523
Availability details for
Quantity
  • Availability
  • Add to Cart