ENA413171908 Enamine

ethyl 2-(adamantan-2-ylidene)-2-cyanoacetate

  • Cas Number : 25694-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156527
Availability details for
Quantity
  • Availability
  • Add to Cart