ENA413171911 Enamine

4-(2,2-difluoroethoxy)-3-methoxybenzaldehyde

  • MDL number MFCD12768595
  • Cas Number : 1179127-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155250
Availability details for
Quantity
  • Availability
  • Add to Cart