ENA413171915 Enamine

1-butyl-4-chloro-3-methyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD15976425
  • Cas Number : 1245772-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155253
Availability details for
Quantity
  • Availability
  • Add to Cart