ENA413171916 Enamine

propane-2-sulfonyl isocyanate

  • MDL number MFCD21879817
  • Cas Number : 89781-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156573
Availability details for
Quantity
  • Availability
  • Add to Cart