ENA413171919 Enamine

2-(propan-2-yloxy)pyridin-4-amine

  • MDL number MFCD16084428
  • Cas Number : 1247371-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156600

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-156600
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart