ENA413171935 Enamine

8-methyl-2,3,4,5-tetrahydro-1,5-benzothiazepine

  • MDL number MFCD27920298
  • Cas Number : 1555384-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156632

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156632
Availability details for
Quantity
  • Availability
  • Add to Cart