ENA413171942 Enamine

2-amino-N,3-diphenylpropanamide hydrochloride

  • MDL number MFCD13562612
  • Cas Number : 857770-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156647

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156647
Availability details for
Quantity
  • Availability
  • Add to Cart