ENA413171952 Enamine

2-[(2-formyl-1H-pyrrol-1-yl)methyl]benzonitrile

  • MDL number MFCD11909539
  • Cas Number : 130569-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156684

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156684
Availability details for
Quantity
  • Availability
  • Add to Cart