ENA413171953 Enamine

2-hydroxy-3-[3-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD21331258
  • Cas Number : 1537294-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156685

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156685
Availability details for
Quantity
  • Availability
  • Add to Cart