ENA413171965 Enamine

7-tert-butyl-1,2,3,4-tetrahydronaphthalen-1-amine

  • MDL number MFCD11591035
  • Cas Number : 1082475-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156723

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156723
Availability details for
Quantity
  • Availability
  • Add to Cart