ENA413171974 Enamine

4-chloro-1,3-dimethyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD15976415
  • Cas Number : 1227241-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155267

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155267
Availability details for
Quantity
  • Availability
  • Add to Cart