ENA413171976 Enamine

tert-butyl N-(2-chlorothiophen-3-yl)carbamate

  • MDL number MFCD27920308
  • Cas Number : 1781900-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156769
Availability details for
Quantity
  • Availability
  • Add to Cart