ENA413171978 Enamine

2-bromo-5-cyclopropyl-1,3,4-oxadiazole

  • MDL number MFCD21881292
  • Cas Number : 1368940-10-6
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-cyclopropyl-1,3,4-oxadiazole
Inchi Code 1S/C5H5BrN2O/c6-5-8-7-4(9-5)3-1-2-3/h3H,1-2H2
Inchi Key CDPWXZZYEQFJFI-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-156772

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-156772
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart