ENA413171979 Enamine

3-(2,2-difluoroethoxy)benzaldehyde

  • MDL number MFCD12807548
  • Cas Number : 467458-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155268
Availability details for
Quantity
  • Availability
  • Add to Cart