ENA413171981 Enamine

[2-chloro-6-(pyrrolidin-1-yl)phenyl]methanol

  • MDL number MFCD10694912
  • Cas Number : 784100-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156809
Availability details for
Quantity
  • Availability
  • Add to Cart