ENA413171983 Enamine

3-(thiophen-3-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16671380
  • Cas Number : 1249642-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156835

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156835
Availability details for
Quantity
  • Availability
  • Add to Cart