ENA413171986 Enamine

[1-(2,2,2-trifluoroethyl)piperidin-4-yl]methanamine

  • MDL number MFCD11131610
  • Cas Number : 335653-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156842

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-156842
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart