ENA413171987 Enamine

3-bromo-2-phenoxypyridine

  • MDL number MFCD14584706
  • Cas Number : 1167991-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156844
Availability details for
Quantity
  • Availability
  • Add to Cart