ENA413171992 Enamine

O-[(2-bromo-4-fluorophenyl)methyl]hydroxylamine

  • MDL number MFCD21921032
  • Cas Number : 1388036-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156855

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156855
Availability details for
Quantity
  • Availability
  • Add to Cart