ENA413172001 Enamine

1-(cyclopropylmethyl)-1H-pyrazole-4-sulfonamide

  • MDL number MFCD24088320
  • Cas Number : 1521489-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156868
Availability details for
Quantity
  • Availability
  • Add to Cart