ENA413172006 Enamine

4-(4-chlorophenyl)-5-methyl-1H-imidazole

  • MDL number MFCD00866069
  • Cas Number : 65571-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155283
Availability details for
Quantity
  • Availability
  • Add to Cart