ENA413172011 Enamine

(2-amino-3-methylbutyl)(methyl)amine

  • MDL number MFCD12796827
  • Cas Number : 1248416-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156911
Availability details for
Quantity
  • Availability
  • Add to Cart