ENA413172012 Enamine

3,3,3-trifluoro-1-(1H-indol-3-yl)propan-1-one

  • MDL number MFCD11211129
  • Cas Number : 1094462-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156912
Availability details for
Quantity
  • Availability
  • Add to Cart