ENA413172016 Enamine

4-methyl-N1-(1-methylpiperidin-4-yl)benzene-1,3-diamine

  • MDL number MFCD17247977
  • Cas Number : 1338984-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156926
Availability details for
Quantity
  • Availability
  • Add to Cart