ENA413172017 Enamine

(4-chloro-2-fluorophenyl)methanamine hydrochloride

  • MDL number MFCD00143286
  • Cas Number : 202982-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 243...245
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155286

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-155286
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart