ENA413172020 Enamine

5-(pyrrolidin-2-yl)thiophene-2-carboxylic acid hydrochloride

  • MDL number MFCD13186026
  • Cas Number : 1269054-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156939
Availability details for
Quantity
  • Availability
  • Add to Cart