ENA413172022 Enamine

2-(6-amino-2-oxo-1,2,3,4-tetrahydroquinolin-1-yl)acetonitrile

  • MDL number MFCD12172564
  • Cas Number : 1156903-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156942
Availability details for
Quantity
  • Availability
  • Add to Cart