ENA413172023 Enamine

5-amino-2-{[2-(pyridin-3-yl)ethyl]amino}benzoic acid

  • MDL number MFCD12895812
  • Cas Number : 1183838-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156946
Availability details for
Quantity
  • Availability
  • Add to Cart