ENA413172035 Enamine

2-chloro-1-(2,2-dimethylpyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD18295230
  • Cas Number : 919111-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156972

Safety Information

msds https://www.enaminestore.com/catalog/EN300-156972
Availability details for
Quantity
  • Availability
  • Add to Cart