ENA413172036 Enamine

1-[(2-phenyl-1,3-oxazol-4-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD11600629
  • Cas Number : 1177346-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-156973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156973
Availability details for
Quantity
  • Availability
  • Add to Cart