ENA413172048 Enamine

3,3,3-trifluoro-1-[4-(trifluoromethyl)phenyl]propan-1-amine

  • MDL number MFCD17126302
  • Cas Number : 1270476-37-3
  • Molecular Weight : 257.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-157024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157024
Availability details for
Quantity
  • Availability
  • Add to Cart