ENA413172053 Enamine

tert-butyl N-{[(prop-2-yn-1-yl)carbamoyl]methyl}carbamate

  • MDL number MFCD24391812
  • Cas Number : 847490-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157029
Availability details for
Quantity
  • Availability
  • Add to Cart