ENA413172054 Enamine

2-bromo-5-(1,2-oxazol-3-yl)-1,3,4-thiadiazole

  • MDL number MFCD18345202
  • Cas Number : 1342234-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157030
Availability details for
Quantity
  • Availability
  • Add to Cart