ENA413172061 Enamine

1-bromo-4,4-dimethylpentane

  • MDL number MFCD02258466
  • Cas Number : 6570-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157041

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P270,P271,P280,P301+P312,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P370+P378,P403+P235,P501
MSDS https://www.enaminestore.com/catalog/EN300-157041
pictograms GHS02,GHS05,GHS07
hstatements H226,H302,H312,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart