ENA413172064 Enamine

3-bromo-2-methyl-5-(trifluoromethyl)pyridine

  • MDL number MFCD13193665
  • Cas Number : 1211542-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157084
Availability details for
Quantity
  • Availability
  • Add to Cart