ENA413172065 Enamine

2,4-dichloro-5,6-dimethylpyrimidine

  • MDL number MFCD09910263
  • Cas Number : 1780-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155311

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155311
Availability details for
Quantity
  • Availability
  • Add to Cart