ENA413172070 Enamine

[4-(bromomethyl)cyclohexyl]benzene

  • MDL number MFCD21776162
  • Cas Number : 1521961-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157118
Availability details for
Quantity
  • Availability
  • Add to Cart