ENA413172071 Enamine

3-benzylcyclopentan-1-amine

  • MDL number MFCD20313379
  • Cas Number : 1409471-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157119
Availability details for
Quantity
  • Availability
  • Add to Cart