ENA413172076 Enamine

1,3-dimethyl-5-(3-methyl-1H-pyrazol-1-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD16778542
  • Cas Number : 1251301-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157136

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157136
Availability details for
Quantity
  • Availability
  • Add to Cart