ENA413172078 Enamine

1-[2-(trifluoromethyl)phenyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD07374436
  • Cas Number : 886366-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157138

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157138
Availability details for
Quantity
  • Availability
  • Add to Cart