ENA413172079 Enamine

7-chloro-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD11590868
  • Cas Number : 69739-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155316

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155316
Availability details for
Quantity
  • Availability
  • Add to Cart