ENA413172080 Enamine

tert-butyl 5-bromo-2-methylbenzoate

  • MDL number MFCD21508504
  • Cas Number : 1202551-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157140
Availability details for
Quantity
  • Availability
  • Add to Cart