ENA413172081 Enamine

N-[(2-chlorophenyl)methyl]-1-cyanocyclopropane-1-carboxamide

  • MDL number MFCD23444863
  • Cas Number : 1517663-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157141
Availability details for
Quantity
  • Availability
  • Add to Cart