ENA413172082 Enamine

4-hydroxyquinazoline-5-carbonitrile

  • MDL number MFCD15527379
  • Cas Number : 1260658-66-9
  • Molecular Weight : 171.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxyquinazoline-5-carbonitrile
Inchi Code 1S/C9H5N3O/c10-4-6-2-1-3-7-8(6)9(13)12-5-11-7/h1-3,5H,(H,11,12,13)
Inchi Key MPVAOMGGGOLKDL-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-157143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157143
Availability details for
Quantity
  • Availability
  • Add to Cart