ENA413172084 Enamine

5-chloro-2-(cyclohexyloxy)aniline

  • MDL number MFCD07365163
  • Cas Number : 893751-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154993
Availability details for
Quantity
  • Availability
  • Add to Cart